missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Sapacitabine
MedChemExpress Sapacitabine is an orally available nucleoside analog prodrug that is structurally related to cytarabine.
Brand: MedChemExpress HY-16445
This item is not returnable.
View return policy
Specifications
490.64 g/mol | |
Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month | |
98.51% | |
DMSO : 33.33 mg/mL (67.93 mM∣Need ultrasonic) | |
151823-14-2 |
Reference compound | |
Room temperature | |
100 mg | |
Cancer-Kinase/protease | |
CS682∣ CYC682 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction