missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saponin permeating solution, 0.5% w/v soln. in PBS (10X), Thermo Scientific Chemicals
Cell permeabilizing agent
Brand: Thermo Scientific Alfa Aesar J63209.AK
This item is not returnable.
View return policy
Description
Saponin permeating solution, is used as a cell permeabilizing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Saponin permeating solution | |
Difficult to mix. | |
Liquid |
0.5% w/v soln. in PBS (10X) | |
250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction