missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAPS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17819-1-AP-150UL
This item is not returnable.
View return policy
Description
Protein phosphatase regulatory subunits, such as SAPS1, modulate the activity of protein phosphatase catalytic subunits by restricting substrate specificity, recruiting substrates, and determining the intracellular localization of the holoenzyme. SAPS1 is a regulatory subunit for the protein phosphatase-6 catalytic subunit.Specifications
SAPS1 | |
Polyclonal | |
Unconjugated | |
PPP6R1 | |
KIAA1115, PP6R1, PPP6R1, SAP190, SAPS domain family member 1, SAPS domain family, member 1, SAPS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22870, 243819 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TSI3, Q9UPN7 | |
PPP6R1 | |
SAPS1 Fusion Protein Ag12149 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |