Learn More
Sars Envelope Protein Rabbit anti-SARS-CoV, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41062-0.1MG
Product Code. 18193739
  • 529.00 € / 0.10mg
Explore more special offers
Description
Description
Sars Envelope Protein Polyclonal antibody specifically detects Sars Envelope Protein in SARS-CoV samples. It is validated for ELISA
Specifications
Specifications
Sars Envelope Protein
Polyclonal
Unconjugated
PBS
Rabbit
Peptide affinity purified
RUO
1489671
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
ELISA
1 mg/mL
ELISA 1 ug/mL
SARS coronavirus Envelope protein, SARS CoV E protein, SARS CoV Envelope protein, SARSCoV, SARSCoV Envelope protein, Severe acute respiratory syndrome
Anti-Sars Envelope Protein antibody was raised against a peptide corresponding to 11 amino acids near the carboxy terminus of SARS-CoV Envelope protein. The immunogen is located within the last 50 amino acids of SARS Envelope Proteins.
0.1 mg
Primary
SARS-CoV
Purified