missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Sartobind™ Rapid A Lab
Protein A affinity chromatography unit offering a fast, flexible and future-proof solution for antibody purification.
154.00 € - 528.00 €
Specifications
Flow Rate | 2.5 to 25 mL/min. |
---|---|
Column Type | Affinity |
Stationary Phase | Agarose or Protein A |
Diameter (Metric) Inner Column | 25 mm |
Volume (Metric) | 0.5 mL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30178672
|
Sartorius
SBLRA025EL-1 |
1 Unit |
154.00 €
1 pack |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30178671
|
Sartorius
SBLRA025EL-A |
4 Units |
528.00 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- High flow rates and reduced residence times take you from equilibration to pure antibody in less than two minutes.
- Maximum dynamic binding capacity supports the preparation of your antibody sample to the highest possible yield and purity.
- Ready-for-use units avoid the need for column packing and buffer degassing, while enabling convenient in-line prefiltration.
- Enhanced pressure resistance in a platform that ensures reliable purification with or without a liquid chromatography system.
- Future-proof by design, ensuring ease-of-transfer for your most promising targets to clinical and commercial production.
Specifications
2.5 to 25 mL/min. | |
Agarose or Protein A | |
0.5 mL |
Affinity | |
25 mm |