missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Sartolab™ RF Vacuum Filter
Sartolab RF are disposable vacuum filtration units.
Brand: Sartorius 180A1----------E
This item is not returnable.
View return policy
Description
The units including receiver flask (RF) contain an optimized Sartorius SFCA membrane and combine highest flow-rates and throughput with extremely low protein binding and extractables. They are therefore the product of choice for particle removal from tissue culture media, buffers, additives and other aqueous biologicals. Sartolab RF vacuum units are available as 250 ml, 500 ml and 1000 ml units.
- Low protein binding
- Fast flow and high throughput
- Easy to use
- Including receiver flask
Additional Specifications:
- Max Temperature: 70°F
- Housing Material: Polystyrene (PS)
- Cytotoxicity: USP Biological Reactivity Tests, in vitro <87>
- PH Range: 4.0-8.0
- Sterilization Method: Gamma Radiation
Specifications
Clear/Yellow | |
Surfactant-free Cellulose Acetate | |
24 cm2 | |
250 mL |
Plug-on Hose Barb | |
0.45 μm | |
Filtration Device | |
49 x 49mm |