missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartolab™ RF Vacuum Filtration Unit
BIOPHARMAGREENER_CHOICE

Sartorius Sartolab™ RF vacuum filtration units are single-use units for the sterile filtration of a sample.

Brand:  Sartorius 180F03---------E

181.45 EUR valid until 2024-12-31
Use promo code "21897" to get your promotional price.


 View more versions of this product

Product Code. 17454422

  • 185.00 € / Pack of 12
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Comprised of:

  • A graduated funnel with a polyethersulfone (PES) membrane, a vacuum adapter and a lid
  • A bottle, with graduation and writing field
  • A tube connector for vacuum connection (for stand-alone filtration)
  • A screw cap for storage of the filtrate (individually wrapped to maintain sterility)

Benefits:

  • Highest flow rates with a large filtration area
  • No loss of protein with a low protein affinity membrane
  • Low hold-up volume due to an optimized membrane support
  • Can be used as stand-alone system or with the Sartolab™ Multistation, which allows parallel filtration of up to 6 samples
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
Polystyrene
0.45 μm
Sterile
Sartolab RF
PES
250 mL
Receiver Flask
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers