Learn More
Sartorius Sartolab™ Single-Use Polystyrene Storage Bottle
BIOPHARMAGREENER_CHOICE
Sartorius Sartolab single-use polystyrene bottles for storage of media, buffers, and other aqueous solutions.
74.15 € - 148.00 €
Specifications
Disposable Single-use
For Use With (Equipment) Sartolab
Material Polystyrene
Material (Membrane) Polystyrene
Sterility Sterile
View More Specs
Products 4
Product Code Brand Sample Volume Price Quantity & Availability  
Product Code Brand Sample Volume Price Quantity & Availability  
17404412
BIOPHARMAGREENER_CHOICE
View Documents
Sartorius
180-22---------E
150 mL
74.15 €
Pack of 12
In Stock
to see stock.
Add to basket
17414412
BIOPHARMAGREENER_CHOICE
View Documents
Sartorius
180-23---------E
250 mL
92.60 €
Pack of 12
In Stock
to see stock.
Add to basket
17424412
BIOPHARMAGREENER_CHOICE
View Documents
Sartorius
180-24---------E
500 mL
112.00 €
Pack of 12
In Stock
to see stock.
Add to basket
17434412
BIOPHARMAGREENER_CHOICE
View Documents
Sartorius
180-25---------E
1000 mL
148.00 €
Pack of 12
In Stock
to see stock.
Add to basket
Description
Description
  • The bottles can be used with all Sartolab™ RF | BT vacuum filtration units with a 45 mm neck size
Specifications
Specifications
Single-use
Polystyrene
Sterile
Sartolab
Polystyrene