missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Accessories for Combisart™ Systems: Frits

Frits for use with Sartorius Combisart Systems

163.00 €

Specifications

Product Type Frit
Material Stainless Steel
Quantity 1 Ea.
For Use With (Application) Ensure uniform distribution of microorganisms and particles on the membrane filter surface
For Use With (Equipment) Stainless Steel manifold
View More Specs

Products 2
Product Code Brand Diameter (English) Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (English) Diameter (Metric) Price Quantity & Availability  
10374901
View Documents
Sartorius
6980102
1.9 in. 5cm
163.00 €
Each
In Stock
to see stock.
Add to basket
 
10727622
View accessory products View Documents
Sartorius
6980103
1.8 in. 4.7cm
163.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • Stainless steel
  • For stainless steel manifolds
  • Choose from 47mm and 50mm

Specifications

Specifications

Frit
1 Ea.
Stainless Steel manifold
Stainless Steel
Ensure uniform distribution of microorganisms and particles on the membrane filter surface
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.