Learn More
Sartorius Accessories for the MD8 Airscan™ Air Sampler
Accessories for use with the MD8 Airscan™ Air Sampler. The MD8 is used for quantitative detection of airborne organisms, mainly in sterile areas of Class A/B cleanrooms, isolators, or blow-fill-seal machines.
Brand:  Sartorius 17803
 View more versions of this product
Product Code. 15952590
  • 212.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Accessories
Adapter, Connector - 14-555-869
  • Connects airscan to isolator via flexible PVC hose and filter capsule
  • DN 30 hose barb to 1 to 11 in. sanitary flange
Adapter, Connector - 14-555-890
  • Connects airscan to isolator via silicone tubing and filter capsule
  • DN 25 hose barb to 1 to 11 in. sanitary flange
Adapter, Plate - 14-555-876
  • For culture media plates
Adapter, Ring - 14-555-912
  • Replacement part for aluminum stack
  • Intermediate (middle) part
Aluminum Stack - 14-555-878
  • Stack of 10 open filter holders
Calibration Kit - 14-555-868
  • Portable flowrate measuring device for the MD8 series air samplers
Capsule, Sartoflor - 14-555-889
  • With PTFE membrane and sanitary flange inlet and outlet
  • For sterile air filtration inserted between the MD8 airscan and isolator
Carrying Case - 14-555-873
  • Stable case for the transport and the storage of unit and accessories
Clamp, Flange - 14-555-891
  • For 1 to 11 in. sanitary flanges
Connector - 14-555-879
  • Aluminum
  • Set of connectors for MD 8 (SM 16743/44)
Connector - 14-555-909
  • Aluminum
  • Connects air sampler inlet to flexible hose
Connector - 14-555-910
  • Aluminum
  • Connects flexible hose to filter holder/adapter
Connector - 14-555-911
  • Flexible hose to filter holder/adapter (S-F)
  • For triclamp
Filter Holder - 14-555-880
  • Holds 80 mm gelatin filters
  • Replacement part for the aluminum stack
Filter Holder - 14-555-884
  • Adapter for gelatin filter units (gelatin disposables) to fit on the MD8 inlet
O-Ring - 14-555-922
  • 7.5 x 1.5 mm
Remote Control - 14-555-877
  • For use with PC (dialog system software)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reusable
MD8 Airscan Air Sampler
Culture Media Plates
Adapter