missing translation for 'onlineSavingsMsg'
Learn More

Sartorius arium™ UV Lamp

Specially designed for arium™ pro 611UV and 611VF laboratory water purifications systems

Brand:  Sartorius 611CEL1

457.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10613990

  • 469.00 € / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Dual UV radiation at 254 nm and 184 nm wavelengths
  • Used as an bactericide or organic compound breakdown tool
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Arium Water Purification System
Lamp
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius arium™ UV Lamp > For 611 and pro, UV and VF laboratory water purification systems

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.