missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Arium™ Water Guard

Arium™ Water Guard detects water leaks early. That means your lab gets optimal protection against water damage. Thanks to its high-grade materials and its finely-tuned sensors, it is the ideal partner for laboratory-grade and ultrapure water systems.

Brand:  Sartorius 610AWG1

506.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11790136

  • 523.00 € / Each
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Arium™ Water Guard detects water leaks early. That means your lab gets optimal protection against water damage. Thanks to its high-grade materials and its finely-tuned sensors, Arium™ Water Guard is the ideal partner for laboratory-grade and ultrapure water systems. Leaks are registered by an optical sensor. This automatically closes the inlet line and triggers an acoustic warning signal. In addition, the user can also constantly monitor the system status on the built-in LED display.

  • Reliably protects against leaks
  • Highly sensitive, also suitable for ultrapure water
  • High-grade material, non-corrosive
  • Optic and acoustic alarm
  • Easy to install
  • Integrated wall holder for solenoid valve
  • Automatically blocks the feed water line when a leak is detected
  • Electrically powered
  • Versatile
  • Areas of application: For installation in feed water inlets on Arium™ systems
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Arium Water Guard
41°F to 86°F
Arium™ Systems
For installation in feed water inlets on Arium™ systems
5°C to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Arium™ Water Guard > Arium™ Water Guard; For use with: Arium™ Systems

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.