Learn More
Sartorius Cubis™ Essential Semimicro Balances
The right choice for users without complex operations who primarily want to perform ultra-precise weighing. Sartorius Cubis Essential Semimicro Balances have robust, monolithic weighing system technology for reliable measuring results.
Brand:  Sartorius MSE225S-ED15
Product Code. 15334278
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Features
  • Large, high-contrast, liquid crystal Cubis MSE display
  • Easy-to-understand menu guidance with short text prompts
  • Clearly structured keys for precise activation of functions
  • Software support for fast leveling
  • Easy calibration with isoCAL (internal automatic calibration | adjustment function)
  • Amply dimensioned, rectangular weighing pans
  • Data Interfaces: USB and RS232C; 3 optional ports are available as an accessory (Bluetooth, PS/2, RS232C)
  • Choose from 13 preconfigured premium Cubis Lab Balances, including semi-micro, analytical, precision and high-capacity balances
Draft Shields
  • Semi-micro and analytical models (14-559-000, 001, 002) include draft shields with high mechanical stability and effortlessly silent moving doors.
  • No disturbing frame, for unhindered access to the weighing pan.
  • Protected against electrostatic influences by a conductive coating
Applications
Unit conversion, isoCAL automatic calibration/adjustment function, density determination (buoyancy method only), calculations, averaging, net/total formulation, weighing in percent, counting,
totalizing.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.01 mg
≤0.1 mg
0 to 60 g: 0.015 mg; 60 to 220 g: 0.025 mg
85 mm
Slotted weighing pan; separate electronics box
MSE225S-ED15
≤6 sec.
85 mm