Learn More
Sartorius Entris™ Precision Balances
Delivers effective and reliable weighing results in daily work. Sartorius Entris™ Precision Balances provide easy and clearly structured user interface, logical key assignment and excellent readability for error-free operation.
1000.00 € - 1760.00 €
Specifications
Readability 1 mg
Linearity 2 mg
Repeatability 1 mg
Dimensions (D x W x H) 230 x 303 x 136 mm
Diameter (Metric) Outer Pan 115 mm
View More Specs
Products 8
Product Code Brand Capacity Model Stabilization Time Price Quantity & Availability  
Product Code Brand Capacity Model Stabilization Time Price Quantity & Availability  
15263913
View Documents
Sartorius
ENTRIS623-1S
620 g ENTRIS623-1S 1.0 sec. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15283913
View Documents
Sartorius
ENTRIS153-1S
150 g ENTRIS153-1S 1.3 sec. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15273913
View Documents
Sartorius
ENTRIS423-1S
420 g ENTRIS423-1S 1.0 sec. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15293913
View Documents
Sartorius
ENTRIS323-1S
320 g ENTRIS323-1S 1.1 sec. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15253903
View Documents
Sartorius
ENTRIS323I-1S
320 g ENTRIS323i-1S 1.1 sec. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15223903
View Documents
Sartorius
ENTRIS423I-1S
420 g ENTRIS423i-1S 1.0 sec. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15233903
View Documents
Sartorius
ENTRIS153I-1S
150 g ENTRIS153i-1S 1.3 sec. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15203903
View Documents
Sartorius
ENTRIS623I-1S
620 g ENTRIS623i-1S 1.0 sec. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
  • New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Specifications
Specifications
1 mg
1 mg
115 mm
2 mg
230 x 303 x 136 mm