Learn More
Sartorius Replacement parts for Microsart Funnel
Sartorius Microsart Funnels are ready-to-use, sterile, polypropylene funnels with click-fit closure.
Brand:  Sartorius 1ZU---0002
151.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11723859
  • 200.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 14-555-783: Microsart™ Frit
  • 14-555-784: Microsart Base 47 mm, stainless steel
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Microsart Funnel
Each
4.7 cm
Stainless Steel
Microsart Base Support