missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Vivaflow™ 200
BIOPHARMA

Laboratory cross flow cassette can process up to 5 liters for cell culture supernatant and other laboratory applications

Brand:  Sartorius VF20P9

514.85 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


Product Code. 15903040

  • 730.00 € / Each
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket

Includes:
  • Has all needed accessories for operating with laboratory pump
This item is not returnable. View return policy

Description

Description

This reusable unit can be used alongside a second cross flow cassette.

  • Reusable
  • Polyethersulfone membrane
  • 500 to 5000mL (17 to 170 oz.) volume range
  • 3000 Dalton MWCO
  • Can be used with another unit simultaneously
  • Cell culture supernatant
  • Virus concentration
  • Desalting
  • Buffer exchange
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Centrivap
Crossflow Cassette
5 L
4.96 in.
0.4 cm
Acrylic Housing and Flow Channel, Polypropylene Membrane Support, Silicone Seals and O-ring
0.39 in.
Polypropylene and Stainless Steel spring pressure indicator, Polypropylene flow restrictor and size 16 PVC peristaltic tubing and Nylon fittings
169.07 oz.
60°C
5.43 in.
12.6 cm
1 cm
4.96 x 0.39 x 5.43 in. (12.6 x 1 x 0.4 cm)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number243500377 Certificate TypeCertificate of Quality Date20/03/2025 Catalog NumberVF20P9
Lot Number240900717 Certificate TypeCertificate of Quality Date25/06/2024 Catalog NumberVF20P9
Lot Number240700817 Certificate TypeCertificate of Quality Date25/06/2024 Catalog NumberVF20P9
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Stedim Vivaflow™ 200 > Module, Concentrator

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.