missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaflow™ 50R Crossflow Casettes
BIOPHARMA

Unique, Plug and Play laboratory crossflow devices for concentrating samples like viruses and antibodies

Brand:  Sartorius VF05H1

307.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15730258

  • 396.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings
This item is not returnable. View return policy

Description

Description

  • Reusable device with low-binding Hydrosart™ (modified regenerated cellulose) membrane
  • For concentrating up to 1L samples
  • Active membrane area per device is 50cm2
  • One unit comes with all necessary accessories for running device with laboratory pump and size 16 pump head
  • For speeding up concentration, two cassettes can be run simultaniously
  • Acrylic main housing and flow channel
  • Polypropylene membrane support
  • Nylon fittings and PVC tubing
  • Compatible with 1M NaOH for cleaning and depyrogenation

  • High flow rate and recovery
  • L x W x H: 100x 24 x 100mm
  • Pump flow rate: 200 to 400mL/min
  • Maximum pressure: 45psi
  • Maximum temperature: 60°C
    • Antibody/recombinant protein concentration/diafiltration in biopharma research
    • Concentration of small production lots of proteins for diagnostic purposes
    • Concentration of viruses from cell culture supernatants or of environmental samples
    • Nanoparticle concentration

    TRUSTED_SUSTAINABILITY
Specifications

Specifications

Acrylic
Concentrating viruses and antibodies
Protein, Virus, Nanoparticles
5 kDa
Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings
100 mL to 1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.