missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivaspin™ 2 Centrifugal Concentrator with Hydrosart™ Regenerated Cellulose (RC) Membrane
BIOPHARMA

Vivaspin™ 2 Centrifugal Concentrators are disposable ultrafiltration units for concentration of macromolecules. With a broad range of molecular weight cut-offs (MWCO) and membrane materials they are suitable for various substrates, including proteins, viruses, nucleic acids, and nanoparticles.

232.00 € - 780.00 €

Specifications

Disposable Single-use
Filter Type Membrane Filter
For Use With (Equipment) Swinging Bucket Rotor (3 mL), Fixed-angle Rotor (2 mL)
Material (Membrane) Hydrosart
Sample Volume 3 mL
View More Specs

Products 8
Product Code Brand MWCO Quantity Price Quantity & Availability  
Product Code Brand MWCO Quantity Price Quantity & Availability  
11778813
BIOPHARMA
View Documents
Sartorius™
VS02H01
10 kDa 25 Pk.
232.00 €
Pack of 25
In Stock
to see stock.
Add to basket
 
11770726
BIOPHARMA
View Documents
Sartorius™
VS02H02
10 kDa 100 Pk.
780.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
11768813
BIOPHARMA
View Documents
Sartorius™
5 kDa 25 Pk.
232.00 €
Pack of 25
In Stock
to see stock.
Add to basket
 
11780726
BIOPHARMA
View Documents
Sartorius™
VS02H12
5 kDa 100 Pk.
780.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
11788813
BIOPHARMA
View Documents
Sartorius™
VS02H21
30 kDa 25 Pk.
232.00 €
Pack of 25
In Stock
to see stock.
Add to basket
 
11877493
BIOPHARMA
View Documents
Sartorius™
VS02H91
2 kDa 25 Pk.
232.00 €
Pack of 25
In Stock
to see stock.
Add to basket
 
11700736
BIOPHARMA
View Documents
Sartorius™
2 kDa 100 Pk.
780.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
11790726
BIOPHARMA
View Documents
Sartorius™
VS02H22
30 kDa 100 Pk.
780.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
Description

Description

Vivaspin concentrators are designed for the concentration or diafiltration of initial sample volumes up to 3 mL and can be used with swinging bucket or fixed angle rotors that accept 15 mL centrifuge tubes. Vivaspin's patented vertical membrane design and thin channel filtration chamber (US 5,647,990) minimize membrane fouling and provide high-speed concentration for even particle-laden solutions. The speed of Vivaspin products combined with low internal surface and membrane area allow superior recovery from very dilute solutions. The Vivaspin 2 also offers flexibility for process optimization: one can directly pipette the concentrate from the dead stop pocket (built into the bottom of the concentrator) or reverse-spin the concentrate into the recovery cap (which can be sealed for storage). Either method results in near-total concentrate recovery.

  • High retentate recovery >90%
  • Vertical membrane design for ultra fast concentration
  • Universal rotor compatibility
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Optional re-spin for concentrate recovery
  • Volume graduation for easy reading/monitoring

Specifications

Specifications

Single-use
Swinging Bucket Rotor (3 mL), Fixed-angle Rotor (2 mL)
3 mL
Centrifugal Concentrator
126 mm
Membrane Filter
Hydrosart
Non-sterile
5 in.
Videos
SDS
Documents

Documents

Product Certifications