missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane
BIOPHARMA

Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.

Brand:  Sartorius VS0231

 View more versions of this product

Product Code. 10009450

  • 232.00 € / Pack of 25
In Stock
to see stock.
Add to basket
This item is not returnable. View return policy

Description

Description

Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.

Vivaspin concentrators are disposable ultrafiltration devices for concentrating biological samples. Vivaspin 2 is designed for use with samples from 500μl to 2mL in rotors that accept 15mL centrifuge tubes. The unique design provides high-speed concentrations for even particle-laden solutions and near-total concentrate recovery.

Twin vertical membranes and optimal design for fast, safe concentration
  • Patented twin vertical membrane design for ultra-fast processing speeds
  • In a single spin, solution volumes up to 4mL can be concentrated up to 30x in 5–30 minutes
  • Thin-channel filtration chamber minimizes membrane fouling and increases filtration speed
  • Dead stop pocket built into the bottom eliminates the risk of sample filtration to dryness
  • High-flux PES membranes feature very low protein binding and low extractables
Designed for maximum flexibility and sample recovery
  • Low internal surface and membrane area for superior recoveries from very dilute solutions
  • Two methods for easy concentrate recovery:
    • Directly pipet the concentrate from the dead stop pocket in the concentrator
    • Reverse-spin the concentrate into the recovery cap, which can be sealed for storage
    • Both methods provide near-total concentrate recovery of greater than 90%
  • Volume graduation for easy monitoring of concentrate volume and 8μl dead-stop volume
  • Universal compatibility with swing bucket or fixed angle rotors accepting 15mL centrifuge tubes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
50 kDa
Concentration, desalting and buffer applications
Swing bucket rotor: 4,000 x g
Fixed angle rotor: 9,000 x g (3 - 50kDa MWCO); 12,000 x g (100kDa+ MWCO)
2 mL (Fixed Angle Rotor), 3 mL (Swing Bucket Rotor)
17.8 mm
PES
1.3 cm2
Polycarbonate
25/Pk.
126 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers