Learn More
Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane

Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.
Brand: Sartorius VS0291
165.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.
Vivaspin concentrators are disposable ultrafiltration devices for concentrating biological samples. Vivaspin 2 is designed for use with samples from 500μl to 2mL in rotors that accept 15mL centrifuge tubes. The unique design provides high-speed concentrations for even particle-laden solutions and near-total concentrate recovery.
Twin vertical membranes and optimal design for fast, safe concentration
- Patented twin vertical membrane design for ultra-fast processing speeds
- In a single spin, solution volumes up to 4mL can be concentrated up to 30x in 5-30 minutes
- Thin-channel filtration chamber minimizes membrane fouling and increases filtration speed
- Dead stop pocket built into the bottom eliminates the risk of sample filtration to dryness
- High-flux PES membranes feature very low protein binding and low extractables
Designed for maximum flexibility and sample recovery
- Low internal surface and membrane area for superior recoveries from very dilute solutions
- Two methods for easy concentrate recovery:
- Directly pipet the concentrate from the dead stop pocket in the concentrator
- Reverse-spin the concentrate into the recovery cap, which can be sealed for storage
- Both methods provide near-total concentrate recovery of greater than 90%
- Volume graduation for easy monitoring of concentrate volume and 8μl dead-stop volume
- Universal compatibility with swing bucket or fixed angle rotors accepting 15mL centrifuge tubes

Specifications
Single-use | |
Centrifuge | |
3 kDa | |
Non-sterile | |
Concentration, desalting and buffer applications | |
Swing bucket rotor: 4000 x g, Fixed angle rotor: 9000 x g (3 - 50 kDa MWCO); 12,000 x g (100 kDa+ MWCO) | |
Centrifugal Concentrator | |
126 mm |
Ultrafiltration | |
PES | |
0.4 to 3 mL | |
1.3 cm2 | |
Polycarbonate | |
25/Pk. | |
2 mL | |
17.8 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.