missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Sash Cable

Labconco™ Sash Cable is a replacement 130-inch/330cm sash cable for use in compatible Protector™ fume hoods.

Brand:  Labconco™ 4949902

Product Code. 15814868

  • 35.20 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Replacement sash cable: length 130 in./330cm
  • For use with a variety of Protector™ Laboratory Fume Hoods, including:
    • Classmate™
    • Perchloric Acid and Acid Digestion
    • Stainless Steel Radioisotope
    • Premier™ Models
    • XL™ Models
Specifications

Specifications

Protector™ fume hoods
Sash Cable
130 in.
330 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers