Learn More
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Brand:  Thermo Scientific™ RD72
 View more versions of this product
Product Code. 11748609
  • 1075.35 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
72 x 0.5mL microtubes
DNA130
Traditional
Concentrator Rotor
72
0.5 mL