Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Select from one of these rotors to fit the type of sample volume and various sample formats for your specific application. Choose from various size of tubes, flasks, mircrowell plates.
827.12 € - 2321.55 €
Product Code | Brand | Description | Holds | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Holds | For Use With (Equipment) | Price | Quantity & Availability | |||
11758609
|
Thermo Scientific™
RH100-6 |
Glass and Plastic Tubes: 100 (6 x 50mm), Medium capacity models | 100 | SPD1030, SPD120, SPD130, SPD140DDA |
1195.95 €
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||
11949426
|
Thermo Scientific™
RH100-8 |
Glass and Plastic Tubes: 100 (8 x 29mm), Medium capacity models | 100 | SPD1030, SPD120, SPD130, SPD140DDA |
1175.85 €
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||
11738629
|
Thermo Scientific™
RH10-15 |
Centrifuge Tubes: 10 x 15mL conical, Medium capacity models | Centrifuge Tubes | SPD1030, SPD120, SPD130, SPD140DDA |
1477.35 €
Each |
In Stock
Log in to see stock. |
|||
11798539
|
Thermo Scientific™
RH12-20 |
Vials: 12 (20 x 47, 20 x 60mm), Medium capacity models | 12 Vials | SPD1030, SPD111V, SPD121P, SPD140DDA |
1165.80 €
Each |
Estimated Shipment: 01-04-2025
Log in to see stock. |
|||
11768609
|
Thermo Scientific™
RH12-28 |
Vials: 12 (28 x 60mm) scintillation | 28 x 60 mm Scintillation | SPD1030, SPD111V, SPD121P, SPD140DDA |
1175.85 €
Each |
In Stock
Log in to see stock. |
|||
11796427
|
Thermo Scientific™
RH24-15 |
Vials: 24 (1 dram 15 x 45mm, 4mL), Medium capacity models | 24 Vials | SPD1030, SPD111V, SPD121P, SPD140DDA |
1165.80 €
Each |
In Stock
Log in to see stock. |
|||
11798589
|
Thermo Scientific™
RH24-18 |
24-place; 18 x 52mm mini scint vials | 18 x 52 mm Mini-scintillation | SPD1030, SPD120, SPD130, SPD140DDA |
1165.80 €
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||
11768659
|
Thermo Scientific™
RH32-13 |
Glass and Plastic Tubes: 32 (13 x 100mm), Medium capacity models | 32 Glass and Plastic Tubes | SPD1030, SPD120, SPD130, SPD140DDA |
1487.40 €
Each |
Estimated Shipment: 01-04-2025
Log in to see stock. |
|||
11748539
|
Thermo Scientific™
RH40-11 |
Microtubes: 40 x 1.5mL, Medium capacity models | 40 Microtubes | SPD1030, SPD120, SPD130, SPD140DDA |
827.12 €
Each |
In Stock
Log in to see stock. |
|||
11738539
|
Thermo Scientific™
RH40-12 |
Glass and Plastic Tubes: 40 (12 x 75mm), Medium capacity models | 40 Glass and Plastic Tubes | SPD1030, SPD120, SPD130, SPD140DDA |
1165.80 €
Each |
In Stock
Log in to see stock. |
|||
11738619
|
Thermo Scientific™
RH4-100 |
Flasks: 4 x 100mL pear-shaped, Medium capacity models | 4 100 mL Pear-shaped Flask | SPD1030, SPD120, SPD130, SPD140DDA |
2321.55 €
Each |
In Stock
Log in to see stock. |
|||
11708549
|
Thermo Scientific™
RH60-12-40 |
Vials: 60 (12 x 32mm), Medium capacity models | 60 Vials | SPD1030, SPD120, SPD130, SPD140DDA |
1165.80 €
Each |
In Stock
Log in to see stock. |
|||
11718549
|
Thermo Scientific™
RH64-11 |
Microtubes: 64 x 1.5mL, Medium capacity models | 64 Microtubes | SPD1030, SPD120, SPD130, SPD140DDA |
1115.55 €
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||
11728629
|
Thermo Scientific™
RH6-50 |
Centrifuge Tubes: 6 x 50mL conical, Medium capacity models | 6 50 mL Conical | SPD1030, SPD120, SPD130, SPD140DDA |
1477.35 €
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||
11788629
|
Thermo Scientific™
RH72-12 |
Glass and Plastic Tubes: 72 (12 x 75mm), Medium capacity models | 72 Glass and Plastic Tubes | SPD1030, SPD120, SPD130, SPD140DDA |
1467.30 €
Each |
In Stock
Log in to see stock. |
|||
10433332
|
Thermo Scientific™
RH8-18 |
Glass and Plastic Tubes: 8 (18 x 100, 17 x 95, 16 x 100mm), Medium capacity models | 8 | SPD1030, SPD120, SPD130, SPD140DDA |
933.65 €
Each |
Estimated Shipment: 07-04-2025
Log in to see stock. |
|||
Description
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Various size rotors made from either plastic or aluminum to fit various sample formats of your choice.
- Easy to use
- Multi-size tube holes accommodate different sample sizes
- Require no additional accessories
- Fit directly into your concentrator
Specifications
Rotor | |
Traditional |