missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saybolt (ASTM D6045, D156) Colour Reference Standard, Paragon Scientific™
Saybolt Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
181.00 €
Specifications
Quantity | 500 mL |
---|---|
Type | Color Reference Standard |
Certifications/Compliance | Certified according to ISO 17025 / ISO 17034 under UKAS accreditation |
Product Code | Brand | Description | Price | Quantity | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity | Availability | ||||
16080152
|
Paragon Scientific
134040 |
ASTM Color -10 | ASTM Color -10 | N/A | |||||
16090152
|
Paragon Scientific
134050 |
ASTM Color 0 | ASTM Color 0 | N/A | |||||
16000162
|
Paragon Scientific
134060 |
ASTM Color 12 | ASTM Color 12 |
181.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010162
|
Paragon Scientific
134070 |
ASTM Color 25 | ASTM Color 25 | N/A | |||||
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to International Standards
- 500mL Pack Size supplied with MSDS
- 18 months shelf life
- Tamper evident security packaging
- Manufactured in the United kingdom
Specifications
500 mL | |
Color Reference Standard | |
Certified according to ISO 17025 / ISO 17034 under UKAS accreditation |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 500ML Saybolt Colour Reference Standard -10
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Carcinogenicity Category 1B
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : white oil, lubricants
Nominal values quoted
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title