Learn More
Description
SB 225002 is a selective non-peptide CXCR2 inhibitor which suppresses IL-8 induced neutrophil chemotaxis and margination. In addition, SB 225002 reduces MPO activity, IL-1β, MIP-2, and keratinocyte-derived chemokine (KC) levels. Further studies suggest that SB 225002 decreases the expression of vascular endothelial growth factor, inducible NOS (NO synthase), and Cox-2 (cyclooxygenase-2 proteins) in colon tissue.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 5 mg |
Description | Yellow |
Content And Storage | -30°C to -10°C |
For Use With (Application) | A potent, selective, and competitive antagonist of the G-protein coupled receptor CXCR2 |
Formula Weight | 352.10g/mol |
Sensitivity | Moisture Sensitive |
Solubility | Soluble to 100mM in DMSO and to 50mM in ethanol |
Product Type | Signal Transduction Reagent |
Form | Solid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.