missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB 399885 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, brain penetrant, orally active 5-HT6 antagonist. Displays > 200-fold selectivity for 5-HT6 over other 5-HT receptors (pKi values are 9.11, 8.81 and 9.02 for human recombinant, native rat and native human 5-HT6 receptors respectively).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 482.81 |
Percent Purity | >99% |
Chemical Name or Material | SB 399885 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction