Learn More
Tocris Bioscience™ SB 3CT
Selective high affinity MMP2 inhibitor
Brand:  Tocris Bioscience™ 6088/10
253.13 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18755568
  • 320.00 € / 10mg
Explore more special offers
Description
Description
SB 3CT is a selective high affinity MMP2 inhibitor (Ki = 28 nM); also inhibits MMP9 (Ki = 400 nM). Active in vivo, neuroprotective and blood-brain barrier permeable.
Specifications
Specifications
White
292605-14-2
306.4g/mol
Store at -20°C
10 mg
Inhibitor
C15H14O3S2
Soluble to 100mM in DMSO, 20mM in ethanol
Selective high affinity MMP2 inhibitor