Learn More
Tocris Bioscience™ SB 431542, Tocris Bioscience™
Potent, selective inhibitor of TGF-βRI, ALK4 and ALK7
Brand:  Tocris Bioscience™ 1614/50
 View more versions of this product
Product Code. 18064922
  • 1685.00 € / 50mg
Explore more special offers
Description
Description
SB 431542 is a potent and selective inhibitor of the transforming growth factor-β (TGF-β) type I receptor/ALK5 (IC50 = 94 nM), and its relatives ALK4 and ALK7. Suppresses TGF-β-induced proliferation of human osteosarcoma cells. Replaces SOX2 in reprogramming of fibroblasts into iPSCs. Stimulates proliferation, differentiation and sheet formation of ESC-derived endothelial cells. Inhibits TGF-β-induced EMT, migration, invasion and VEGF secretion in several human cancer cell lines.
SB 431542 synthesized to cGMP guidelines also available.
For more information about how SB 431542 may be used, see our protocols: 3D Culture of Lung Alveolar Cells, Differentiation of Hematopoietic Progenitors from hPSCs, Generating Midbrain Dopaminergic Neurons from hPSCs, Generating Vascular Organoids, Accelerated Induction of Cortical Neurons from hiPSCs .
Specifications
Specifications
SB 431542
4-5-benzo 1,3 dioxol-5-yl-4-pyridin-2-yl-1 h-imidazol-2-yl-benzamide;4-4-2 h-1,3-benzodioxol-5-yl-5-pyridin-2-yl-1 h-imidazol-2-yl benzamide
C22H16N4O3
301836-41-9
50 mg
384.39