missing translation for 'onlineSavingsMsg'
Learn More

Scandium arc-cast pellet, 15.9mm (0.63in) dia, Thermo Scientific Chemicals

Product Code. 15442157
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Scandium arc-cast pellet is used in aerospace industry, oil refineries as a tracing agent, and metal halide lamps/light bulbs. It is mainly used for research purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-20-2
Appearance Silver gray pellet
Density 3g/cm
Molecular Formula Sc
MDL Number MFCD00016323
Quantity 1 Pc.
Synonym unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
Solubility Information Reacts with water.
InChI Key SIXSYDAISGFNSX-UHFFFAOYSA-N
SMILES [Sc]
IUPAC Name scandium
Molecular Weight (g/mol) 44.96
PubChem CID 23952
ChEBI CHEBI:33330
Odor Odorless
Form Pieces
Chemical Name or Material Scandium, arc-cast pellet
Show More Show Less
compliance-icons
Product Identifier
  • Scandium arc-cast pellet
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P233-Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Scandium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'