missing translation for 'onlineSavingsMsg'
Learn More

Scandium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

299.00 € - 919.00 €

Chemical Identifiers

CAS 7440-20-2
Molecular Formula Sc
Molecular Weight (g/mol) 44.96
MDL Number MFCD00016323
InChI Key SIXSYDAISGFNSX-UHFFFAOYSA-N
Synonym unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
PubChem CID 23952
ChEBI CHEBI:33330
IUPAC Name scandium
SMILES [Sc]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11369288
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043219.FF
25 x 25 mm
299.00 €
25mm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11316209
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043219.FI
50 x 50 mm
919.00 €
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Chemical Identifiers

7440-20-2
44.96
SIXSYDAISGFNSX-UHFFFAOYSA-N
23952
scandium
Sc
MFCD00016323
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
CHEBI:33330
[Sc]

Specifications

7440-20-2
MFCD00016323
SIXSYDAISGFNSX-UHFFFAOYSA-N
scandium
23952
99.9%
≈0.20g/25x25mm
(REO)
Sc
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
[Sc]
44.96
CHEBI:33330
Odorless
Foil
Scandium foil, 0.1mm (0.004 in.) thick
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III

missing translation for 'einecsNumber' : 231-129-2

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only