missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

113.57 € - 1014.05 €

Chemical Identifiers

CAS 10361-84-9
MDL Number MFCD00011221
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11323457
SDS View Documents
Thermo Scientific Alfa Aesar
018677.01
0.25 g
113.57 €
0.25g
Please to purchase this item. Need a web account? Register with us today!
11343457
SDS View Documents
Thermo Scientific Alfa Aesar
018677.03
1 g
308.54 €
1g
Please to purchase this item. Need a web account? Register with us today!
11333457
SDS View Documents
Thermo Scientific Alfa Aesar
018677.06
5 g
1014.05 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Scandium(III) chloride is a Prins cyclization catalyst. Used to form an intermetallic nickel-scandium compound film by potentiostatic cathodic reduction from molten salt mixtures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10361-84-9
MFCD00011221

Specifications

939°C
MFCD00011221
Soluble in water; Insoluble in alcohol
99.9%
Hygroscopic
(REO)
10361-84-9
14,8392
151.32
Odorless
2.39 g/mL
Scandium(III) chloride, Anhydrous
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 233-799-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.