missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) chloride hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

120.60 € - 423.11 €

Chemical Identifiers

CAS 20662-14-0
Molecular Formula Cl3H12O6Sc
Molecular Weight (g/mol) 259.40
MDL Number MFCD00149852
InChI Key FKZFOHABAHJDIK-UHFFFAOYSA-K
IUPAC Name scandium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Sc+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326268
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011218.03
1 g
120.60 €
1g
Please to purchase this item. Need a web account? Register with us today!
11316268
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011218.06
5 g
423.11 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Scandium(III) chloride hexahydrate, which is freely soluble in water is used as a source for producing metallic scandium by electrolysis. It finds use in halide lamps, optical fibers, electronic ceramics and lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20662-14-0
259.40
FKZFOHABAHJDIK-UHFFFAOYSA-K
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Sc+3]
Cl3H12O6Sc
MFCD00149852
scandium(3+) hexahydrate trichloride

Specifications

20662-14-0
Cl3H12O6Sc
14,8392
FKZFOHABAHJDIK-UHFFFAOYSA-K
scandium(3+) hexahydrate trichloride
259.41 (151.32 Anhydrous)
Reacton™
(REO)
Crystalline Aggregates
MFCD00149852
Soluble in water.
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Sc+3]
259.40
99.9%
Hygroscopic
Scandium(III) chloride hexahydrate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 233-799-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.