Learn More
Scandium(III) nitrate hydrate, REacton™, 99.99% (REO)
Description
Scandium(III) nitrate hydrate is used as an oxidizing agent in organic synthesis. It is used in the optical coatings, electronic ceramics and laser industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Crystalline Aggregates |
| Quantity | 5 g |
| UN Number | UN1477 |
| Merck Index | 14,8392 |
| Solubility Information | Soluble in water and alcohol. |
| Formula Weight | 230.97 (Anhydrous) |
| Percent Purity | 99.99% |
| Grade | Reacton |
| Sensitivity | Hygroscopic |
| Assay | (REO) |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.