missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scandium(III) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals
60.35 € - 203.00 €
Chemical Identifiers
CAS | 12060-08-1 |
---|---|
Molecular Formula | O3Sc2 |
Molecular Weight (g/mol) | 137.91 |
MDL Number | MFCD00011223 |
InChI Key | HJGMWXTVGKLUAQ-UHFFFAOYSA-N |
IUPAC Name | discandium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Sc+3].[Sc+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11396025
|
Thermo Scientific Acros
193960010 |
1 g | Glass bottle |
60.35 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11306035
|
Thermo Scientific Acros
193960050 |
5 g | Glass bottle |
203.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
12060-08-1 | |
137.91 | |
HJGMWXTVGKLUAQ-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Sc+3].[Sc+3] |
O3Sc2 | |
MFCD00011223 | |
discandium(3+) trioxidandiide |
Specifications
>1000.0°C | |
White | |
99.999% | |
SC2O3 | |
10,8239 | |
HJGMWXTVGKLUAQ-UHFFFAOYSA-N | |
discandium(3+) trioxidandiide | |
137.91 | |
Trace Metal Basis |
12060-08-1 | |
Powder | |
O3Sc2 | |
MFCD00011223 | |
Solubility in water: insoluble | |
[O--].[O--].[O--].[Sc+3].[Sc+3] | |
137.91 | |
99.999% | |
Scandium(III) oxide |
Safety and Handling
missing translation for 'einecsNumber' : 235-042-
RUO – Research Use Only