missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

34.37 € - 326.63 €

Chemical Identifiers

CAS 144026-79-9
Molecular Formula C3F9O9S3Sc
Molecular Weight (g/mol) 492.146
MDL Number MFCD00798539
InChI Key HZXJVDYQRYYYOR-UHFFFAOYSA-K
Synonym scandium iii trifluoromethanesulfonate, scandium trifluoromethanesulfonate, scandium iii triflate, scandium triflate, sc otf 3, scandium 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid scandium iii salt, scandium iii trifluoromethanesulphonate, scandium tris trifluoromethanesulfonate, scandiumtriflate
PubChem CID 2734571
IUPAC Name scandium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Sc+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11400291
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040566.MD
250 mg
34.37 €
250mg
Please to purchase this item. Need a web account? Register with us today!
11318548
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040566.03
1 g
93.77 €
1g
Please to purchase this item. Need a web account? Register with us today!
11328548
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040566.06
5 g
326.63 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Scandium(III) trifluoromethanesulfonate is widely used as a catalyst in hydrothiolation, selective two-electron reduction of oxygen by ferrocene derivatives and vinylogous Fridel-crafts alkylation of indoles and pyrrole in water. It is involved in the Mukaiyama aldol addition and stereochemically catalyzes the radical polymerization of acrylates. It acts as a Lewis acid catalyst and used in the synthesis of bullvalone via a stabilized sulfur ylide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

144026-79-9
492.146
HZXJVDYQRYYYOR-UHFFFAOYSA-K
2734571
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Sc+3]
C3F9O9S3Sc
MFCD00798539
scandium iii trifluoromethanesulfonate, scandium trifluoromethanesulfonate, scandium iii triflate, scandium triflate, sc otf 3, scandium 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid scandium iii salt, scandium iii trifluoromethanesulphonate, scandium tris trifluoromethanesulfonate, scandiumtriflate
scandium(3+);trifluoromethanesulfonate

Specifications

144026-79-9
C3F9O9S3Sc
scandium iii trifluoromethanesulfonate, scandium trifluoromethanesulfonate, scandium iii triflate, scandium triflate, sc otf 3, scandium 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid scandium iii salt, scandium iii trifluoromethanesulphonate, scandium tris trifluoromethanesulfonate, scandiumtriflate
HZXJVDYQRYYYOR-UHFFFAOYSA-K
scandium(3+);trifluoromethanesulfonate
2734571
98%
Scandium(III) trifluoromethanesulfonate
Powder
MFCD00798539
Soluble in water,alcohol and acetonitrile.
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Sc+3]
492.146
492.16
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only