Learn More
Scandium, plasma standard solution, Specpure™ Sc 1000μg/mL, Thermo Scientific Chemicals
Sc 1000μg/mL
113.57 € - 437.18 €
Chemical Identifiers
CAS | 7440-20-2 |
---|---|
Molecular Formula | Sc |
Molecular Weight (g/mol) | 44.96 |
MDL Number | MFCD00016323 |
InChI Key | SIXSYDAISGFNSX-UHFFFAOYSA-N |
PubChem CID | 23952 |
ChEBI | CHEBI:33330 |
IUPAC Name | scandium |
SMILES | [Sc] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338667
|
Thermo Scientific Alfa Aesar
035769.AD |
50 mL |
113.57 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328667
|
Thermo Scientific Alfa Aesar
035769.AE |
100 mL |
202.77 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11455070
|
Thermo Scientific Alfa Aesar
035769.AP |
500 mL |
437.18 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-20-2 | |
44.96 | |
SIXSYDAISGFNSX-UHFFFAOYSA-N | |
CHEBI:33330 | |
[Sc] |
Sc | |
MFCD00016323 | |
23952 | |
scandium |
Specifications
7440-20-2 | |
Sc | |
SIXSYDAISGFNSX-UHFFFAOYSA-N | |
[Sc] | |
44.96 | |
CHEBI:33330 | |
Scandium, plasma standard solution |
23 hPa (17mm Hg) at 20°C | |
MFCD00016323 | |
Liquid | |
scandium | |
23952 | |
Nitric acid: 5%, Scandium(III) oxide: 0.15%, Water: 94.85% |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III
missing translation for 'einecsNumber' : 231-714-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.