Learn More
Merck Scepter™ 3.0 Handheld Automated Cell Counter Kit
The Scepter™ 3.0 Cell Counter delivers rapid cell counts storing up to 999 counts which can be transferred to Excel.
Brand:  Merck PHCC360KIT
 View more versions of this product
Product Code. 17181731
  • 6880.00 € / 1 set
Explore more special offers
Description
Description
  • For research use only
  • Replaces Scepter™ 2.0 and not compatible with 2.0 sensors
  • Provides fast, convenient method for counting cells and particles
  • Compatible with both the 40 and 60 μM sensors.
  • Employs the Coulter Principle in a miniaturized, handheld, format, taking less than 30 seconds
  • Prepare a dilution of the cell culture of interest and use the Scepter™ cell counter to aspirate a sample of this dilution
  •  Displays:
    • Cell concentration
    • Average cell size
    • Average cell volume
    • Histogram of size or volume distribution
TRUSTED_SUSTAINABILITY
Specifications
Specifications
60 μm Scepter 3.0 Cell Counter and Sensors Kit
8 to 25 μm
Cell Counting
Scepter 3.0 Cell Counter, Test Bead Vial, Scepter 3.0 Charging Station and Pk. of Scepter 3.0 Sensors
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Merck Scepter™ 3.0 Handheld Automated Cell Counter Kit > 60 μm Cell Counter Sensors