Learn More
BRAND™ Schilling Burette Bottle
Schilling Burette Bottle equipped with screw cap and tube bushing.
Brand:  BRAND™ 130260
8.88 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 10115243
  • 10.05 € / Each
Estimated Shipment: 05-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Constructed of low density polyethylene
  • Narrow-mouth
  • With tube bushing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw cap
Shilling Burette
Screw cap and tube brushing
Amber
LDPE