missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente™ Schlenk tubes with NS Stopcock
Nitrogen tubes (Schlenk tubes) with conical ground joint and NS stopcock
Brand: Lenz Laborglasintrumente™ 03370228
This item is not returnable.
View return policy
Description
- With conical ground joint and NS stopcock
- Material: DURAN™
Specifications
50 mL | |
Duran Glass | |
Center neck - Cone 14/23, Valve bore - 2.5 mm | |
1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction