Learn More
Eppendorf™ Lab Shaker Clamps
Unlock the potential of your experiments with Eppendorf's versatile range of shaker clamps. Securely fasten Erlenmeyer flasks, Thomson Ultra Yield™, Optimum Growth™, Tunair™, Corning™, and more to your universal platforms. Find the ideal clamp for every flask type.
25.35 € - 143.00 €
Product Code | Brand | Holds | Dimensions (L x W) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Dimensions (L x W) | Price | Quantity & Availability | ||||
10706164
|
Eppendorf™
ACE-10S |
10 mL Erlenmeyer Flasks | 31 x 26 mm |
25.35 €
Each |
In Stock
Log in to see stock. |
||||
10487651
|
Eppendorf™
M1190-9004 |
25 mL Erlenmeyer Flasks | 23 x 22 mm |
25.50 €
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
||||
11386034
|
Eppendorf™
M1190-9000 |
50 mL Erlenmeyer Flasks | 51 x 26 mm |
26.80 €
Each |
In Stock
Log in to see stock. |
||||
11396034
|
Eppendorf™
M1190-9001 |
125 mL Erlenmeyer Flasks | 68 x 37 mm |
28.25 €
Each |
In Stock
Log in to see stock. |
||||
11306044
|
Eppendorf™
M1190-9002 |
250 mL Erlenmeyer Flasks | 91 x 47 mm |
40.20 €
Each |
In Stock
Log in to see stock. |
||||
11316044
|
Eppendorf™
M1190-9003 |
500 mL Erlenmeyer Flasks | 102 x 48 mm |
47.45 €
Each |
In Stock
Log in to see stock. |
||||
11399814
|
Eppendorf™
ACSB-500S |
500 mL Media Bottles and TPP Bioreactors 450 mL or 600 mL | 92 x 108 mm |
132.00 €
Each |
In Stock
Log in to see stock. |
||||
11326044
|
Eppendorf™
ACE-1000S |
1 L Erlenmeyer Flasks | 130 x 64 mm |
62.55 €
Each |
In Stock
Log in to see stock. |
||||
11309824
|
Eppendorf™
ACSB-1000S |
1 L Media Bottles | 108 x 150 mm |
132.00 €
Each |
In Stock
Log in to see stock. |
||||
11698613
|
Eppendorf™
M1190-9005 |
2 L Narrow Neck Erlenmeyer (DIN-ISO 1773), Thomson, or Corning Flasks | 165 x 112 mm |
143.00 €
Each |
In Stock
Log in to see stock. |
||||
11336044
|
Eppendorf™
ACE-2000S |
2 L Wide Neck Erlenmeyer Flasks (DIN-ISO 24450) | 158 x 72 mm |
98.15 €
Each |
In Stock
Log in to see stock. |
||||
11389814
|
Eppendorf™
ACFE-2800S |
2.8 L Fernbach Flasks | 203 x 73 mm |
125.00 €
Each |
In Stock
Log in to see stock. |
||||
11346044
|
Eppendorf™
ACE-3000S |
3 L Erlenmeyer Flasks | 189 x 112 mm |
114.00 €
Each |
In Stock
Log in to see stock. |
||||
11359814
|
Eppendorf™
ACE-4000S |
4 L Erlenmeyer Flasks | 210 x 99 mm |
112.00 €
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
||||
11369814
|
Eppendorf™
ACE-5000S |
5 L Erlenmeyer Flasks | 222 x 145 mm |
121.00 €
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
||||
11379814
|
Eppendorf™
ACE-6000S |
6 L Erlenmeyer Flasks | 244 x 100 mm |
134.00 €
Each |
In Stock
Log in to see stock. |
||||
Description
- Stainless-steel construction (not spot welded or riveted) for added strength and safety.
- Clamps 2 L and above come with an extra girdle for added security when agitating flasks at high speeds.
- Comes complete with required screws, and are fitted with coiled wire springs for added stability at elevated speeds.
Specifications
Clamp | |
Stainless Steel |