missing translation for 'onlineSavingsMsg'
Learn More

SCN4B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55438-1-AP-150UL

 View more versions of this product

Product Code. 16806905

  • 410.00 € / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The SCN4B protein modulates channel gating kinetics. Causes negative shifts in the voltage dependence of activation of certain alpha sodium channels, but does not affect the voltage dependence of inactivation By similarity. The protein has been found to be expressed at a high level in dorsal root ganglia, at a lower level in brain, spinal cord, skeletal muscle and heart. Defects in SCN4B are the cause of long QT syndrome type 10 (LQT10) [MIM:611819]. Long QT syndromes are heart disorders characterized by a prolonged QT interval on the ECG and polymorphic ventricular arrhythmias. They cause syncope and sudden death in response to excercise or emotional stress. They can present with a sentinel event of sudden cardiac death in infancy.
Specifications

Specifications

SCN4B
Polyclonal
Unconjugated
SCN4B
SCN4B, Sodium channel subunit beta 4
Rabbit
Antigen Affinity Chromatography
RUO
399548, 6330
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7M729, Q8IWT1
Scn4b
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.