Learn More
Ansell™ Scorpio™ 8-352 Series Green Neoprene Immersion Gloves
Neoprene dipped coating on interlock knit liner for better grip on wet and slippery materials.
210.00 €
Specifications
Back Material Neoprene
Certifications/Compliance ANSI, EN 388 : 3121, EN 374
Cuff Material Neoprene
Coating Material Neoprene
Laboratory/Cleanroom Use Yes
View More Specs
Products 3
Product Code Brand dimSize Price Quantity & Availability  
Product Code Brand dimSize Price Quantity & Availability  
12649791
View Documents
Ansell™
8352080
210.00 €
12 pairs
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
12639791
View Documents
Ansell™
8352100
210.00 €
12 pairs
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
12659791
View Documents
Ansell™
8352090
210.00 €
12 pairs
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
Description
Description
  • Flexible
  • Textured finish
  • 100% liquidproof
  • Soft stretchy knit liner with no seams in working area
  • Comfortable to wear, cushions the hand, absorbs perspiration and prevents chafing
  • Good wet grip. Safer handling means increased productivity
  • Easier to manipulate, less tiring to the hand and easier to get on and off
  • Neoprene coating
  • Wide ranging chemical protection
  • Certified FDA21 CFR 177-2600
Applications:
  • Cleaning furnaces, distillation pumps, valves or lines and crackers BTX process
  • Equipment repair and maintenance
  • Filling, blending and charging of raw materials
  • Loading and unloading process equipment
  • Opening and draining pumps valves or lines
  • Opening furnaces draining pumps valves or lines and crackers BTX process
  • Transferring liquids and solids
  • Transferring liquids and solids between vessels and tanks and process equipment
Specifications
Specifications
Neoprene
Neoprene
Yes
8-352
Interlock Knit Cotton
72 Pairs
8-352
ANSI, EN 388 : 3121, EN 374
Neoprene
Immersion Gloves
Gauntlet
Fully coated-palm
Chemicals