missing translation for 'onlineSavingsMsg'
Learn More

3M™ Scotch™ C10 Black Tape Dispenser

3M™ Scotch™ C10 Black Tape Dispenser is a sturdy tape rack that fits almost all tape rolls.

Brand:  3M™ 7000080564

Product Code. 15728488

  • 130.00 € / Pack of 6
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sturdy cutter for small and large tape rolls
  • The rack is fitted with a 2.54 cm roll with maximum dimensions of 19 mm x 33 m and a 7.62-centimeter roll with maximum dimensions of 25 mm x 66 m
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tape Dispenser
Core: 1 in. 3 in.
1 Roll up to 66 m x 25 mm (L x W)
Black
Roll of Tape
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Scotch™ C10 Black Tape Dispenser > Color: Black

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.