missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Scott™ Control™ Alcohol Foam Hand Sanitizer

Scott™ CONTROL™ alcohol hand sanitizer kills up to 99.99% of microorganisms. Dermatologically tested and fragrance free, foam sanitizer contains aloe vera and skin humectant.

224.00 € - 230.00 €

Specifications

Product Type Alcohol Foam Hand Sanitizer
Color Clear
Certifications/Compliance (Europe) EN 1500, EN 1276, EN13727, EN 14476, EN 1650, EN 13624, BPR Regulation EU 528/2012
Packaging Cassette
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17163664
View Documents
Kimberly-Clark™
6392
6 x 1 L
224.00 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
17173664
View Documents
Kimberly-Clark™
6393
4 x 1.2 L
230.00 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Features
    • Contains aloe vera and skin humectant designed to support frequent use
    • Dye and fragrance free formulation is dermatologically tested
    • Quick and easy installation with a simple push and click
    • Empty cassette can be compressed to conserve space and is recyclable once pump is removed
  • Uses:
    • Designed to support compliance control in the workplace
    • Used to minimise risk, support hygiene practices and help contamination control in the workplace
  • Notes:
    • Always read label and product information before use
    • Use biocides safely
Specifications

Specifications

Alcohol Foam Hand Sanitizer
Clear
EN 1500, EN 1276, EN13727, EN 14476, EN 1650, EN 13624, BPR Regulation EU 528/2012
Cassette
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ Scott™ Control™ Alcohol Foam Hand Sanitizer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.