missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Hostess™ Essential Couch Cover

Economic and portable, lightweight roll. Hostess™ Essential Couch Cover ideal for protecting couches in the healthcare industry. Strong, tear-resistant and durable.

Brand:  Kimberly-Clark™ 7162

96.96 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 12565518

  • 286.00 € / 6 rolls

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Designed to provide customers with the ideal combination of value and versatility when protecting couches in the healthcare industry
  • The Scott™ Essential Couch Cover (59W) 7162 is soft, absorbent, tear-resistant and durable.
  • These couch covers are delivered in a lightweight, portable roll and are easy to remove and fit.
  • Individual sheet size 37 cm (L) x 59 cm (W).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Couch Cover
Paper
14.6 in.
1-ply
23.2 in.
White
Non-sterile
37 cm
6 Rolls x 198 Sheets
59 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers