missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Scott™ Extra Couch Covers Roll
Scott™ Extra Couch Covers Roll provide quality protection and are ideal for protecting couches in the healthcare industry.
Brand: Kimberly-Clark™ 6024
105.64 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- 2 ply sheets (810 total)
- Ideal for protecting couches and maintaining high levels of hygiene in the healthcare industry
- Quality protection
- Comes in a lightweight and portable roll
Specifications
Couch Cover | |
Paper | |
High Traffic Washrooms | |
6 Rolls | |
2-ply | |
14.6 in. |
Blue | |
59 x 37 cm | |
23.2 in. | |
59 cm | |
6 Pk. | |
37 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction