Learn More
DWK Life Sciences Screw Cap with Aperture, for safety standard ground sockets
For fixing the DURAN™ core in the DURAN™ threaded sleeve.
Brand:  DWK Life Sciences 286215507
 View more versions of this product
Product Code. 15844608
  • 61.50 € / Pack of 10
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The open screw cap reliably fixes the core of the DURAN™ security cuts in the sleeve. When the screw connection is opened, the ground joints are automatically separated from each other.
  • Reliable fixation of the DURAN™ safety sections in one another (core and sleeve)
  • When the screw connection is opened, the ground joints are automatically separated from each other
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
Suitable for fixing DURAN safety grinding
NS 34/35