Learn More
Gosselin™ Screw Cap Dippers with Removable Handle
GREENER_CHOICE
Dip with confidence. Gosselin™ Screw Cap Dippers with Removable Handle prevent spillage with screw caps and detachable shaft.
236.69 € - 796.00 €
Specifications
Material Polypropylene
Sterility Sterile SAL 10-3
Color Blue
View More Specs
Products 3
Product Code Brand Capacity (Metric) Height (Metric) Length (Metric) Handle Price Quantity & Availability  
Product Code Brand Capacity (Metric) Height (Metric) Length (Metric) Handle Price Quantity & Availability  
15488794
GREENER_CHOICE
View Documents
Gosselin™
TAT30C-01
40 mL 80 mm 290 mm including container
796.00 €
Pack of 250
In Stock
to see stock.
Edge
Add to basket
15498794
GREENER_CHOICE
View Documents
Gosselin™
TAT52C-01
125 mL 74 mm 275 mm including container
245.83 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
15408804
GREENER_CHOICE
View Documents
Gosselin™
TAT53C-01
180 mL 102 mm 305 mm including container
236.69 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Polypropylene blue containers
  • Removable handle
  • Shock resistant
  • Packaged individually in tamper evident single zip bags
  • Traceability information on each bag
  • Sterile SAL 10-3
Specifications
Specifications
Polypropylene
Sterile SAL 10-3
Blue