Learn More
Buerkle™ Brass Screw Joint for Barrel Pump
Buerkle™ Brass Screw Joint for Barrel Pump prevents electro-static charge in combination with the anti-static set.
Brand:  Buerkle™ 5601-0100
74.43 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Product Code. 10453481
  • 78.35 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Barrel screw joint made of nickel-plated brass for barrel pump stainless steel
  • Prevents electro-static charge in combination with the Anti-static set
  • For the safe transfer of flammable and easily flammable liquids
  • For containers with R 2 in. steel fine thread inside
  • The container connectors make decanting even easier and safer with their more secure connection to barrels
  • The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange
Specifications
Specifications
Stainless Steel Barrel Pump
Nickel-plated Brass
Barrel Screw Joint