missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Screw Neck Vial, 15-425 thread, Glass, Clear

For storage purposes.

33.25 € - 44.20 €

Specifications

Color Clear
Material Glass
For Use With (Application) Storage
Neck Style Screw
Packaging Type CleanPack, Polypropylene Box
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10504463
View Documents
Fisherbrand™
15 09 1703
8.9 mL
44.20 €
Pack of 100
In Stock
to see stock.
Add to basket
 
11576064
View Documents
Fisherbrand™
15 09 1657
12 mL
33.25 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Constructed from 1st hydrolytic class glass
  • Clear
  • Flat bottom
Specifications

Specifications

Clear
Storage
CleanPack, Polypropylene Box
Flat Bottom
ND15
Glass
Screw
800 μL
100
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.